SuperfleX Flexible Plunger Microliter Syringes, SGE, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The SuperfleX syringe features a super-elastic plunger that returns to its original shape if twisted or bent.
- Syringe volume of 10 μL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.