Autosampler Syringes for Shimadzu AOC 14, 17, 20, and 20i GCs, SGE, Restek
Fournisseur: Restek
Certificats
A propos de cet article
SGE syringes are designed and tested to meet critical autosampler performance needs.
- Compatible with Shimadzu AOC 14, 17, 20, and 20i GCs
Needle point styles are designed to withstand multiple rapid injections through a septum.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.