CarboFrit® Puller/Inserter Tool, Restek
Catalog # RK21642
Supplier: Restek
Specifications
- Description:CarboFrit Puller/Inserter Tool
- Cat. no.:RK21642
- Supplier No.:21642
Specifications
About this item
Hook end for removing CarboFrit® inserts.
- Bent end (90°) for inserting CarboFrit® inserts
- Slender design
- Easy to maneuver
Insert and remove CarboFrit® inserts easily.
Customers Who Bought This Also Bought