GC Installation Kit, Restek
# de catalogue RK21325
Fournisseur: Restek
Spécifications
- Description:GC Installation Kit
- Cat. no.:RK21325
- Supplier No.:21325
Spécifications
A propos de cet article
This kit contains the tubing and fittings needed to add an additional GC to your lab bench.
- Includes everything necessary to installation gas chromatograph
Delivery: Kit includes: tubing cutter, one 1/8 x 1/4" reamer, one 1/4 x 1/8" brass tube end reducer, one 7/16" wrench, one 1/2" wrench, four 1/8" brass tees, ten 1/8" brass nuts, ten brass front and back ferrules, and 50 feet (15.2 meters) of Restek's instrument-grade cleaned 1/8" copper tubing.
Customers Who Bought This Also Bought