Tubing Reamer, Restek
# de catalogue 89205-664
Fournisseur: Restek
Spécifications
- Description:Tubing Reamer
- Cat. no.:89205-664
- Supplier No.:20134
Spécifications
A propos de cet article
Tubing reamers remove burrs from stainless steel tubing.
- For 1/4 or 1/8" tubing
- Nonslip safety design
Customers Who Bought This Also Bought