ContactUsLinkComponent
Tubing Reamer, Restek
Tubing Reamer, Restek
# de catalogue 89205-664
Fournisseur:  Restek
1/8in & 1/4in Tubing Reamer
Tubing Reamer, Restek
# de catalogue 89205-664
Fournisseur:  Restek
Spécifications
  • Description:
    Tubing Reamer
  • Cat. no.:
    89205-664
  • Supplier No.:
    20134
Spécifications
A propos de cet article
Tubing reamers remove burrs from stainless steel tubing.
  • For 1/4 or 1/8" tubing
  • Nonslip safety design
Customers Who Bought This Also Bought