Cylinder Holders, Wall Mounted, Restek
Fournisseur: Restek
Certificats
A propos de cet article
These holders are designed to prevent free-standing gas cylinders from tipping over and injuring personnel.
- Cylinder holders can accommodate 1 to 4 cylinders depending on model
The cast aluminum holder can be secured to a wall or the side of a work bench. Each mount will secure a cylinder 4 to 15 inches in diameter.
Customers Who Bought This Also Bought