Cylinder Valve Wrench, Restek
# de catalogue 10855-832
Fournisseur: Restek
Spécifications
- Description:Cylinder Valve Wrench
- Cat. no.:10855-832
- Supplier No.:21321
Spécifications
A propos de cet article
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel.
It is also suitable for removing difficult cylinder caps.
Customers Who Bought This Also Bought