Hydrogen Flashback Arrestor, Restek
# de catalogue 10855-640
Fournisseur: Restek
Spécifications
- Description:Hydrogen Flashback Arrestor
- Material:Brass Body
- Includes:1/4" female NPT
- Supplier No.:21334
Spécifications
A propos de cet article
Valve cuts off gas supply in the event of a flashback.
- Can be reset and reused after flashback has occurred
- Brass body
- Polyethylene temperature sensor
Certifications: Meet safety requirements of BS 6158, EN 730, ISO 5175, AS 4603 and are UL Listed 23Y5.
Customers Who Bought This Also Bought