Photoionization, PID Lamps, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Multiple models of photoionization detector lamps are available in a variety of sizes and lamp outputs.
Model 108-10.0/10.6 offers both 10.0 and 10.6 eV potential, has a 0.781" base diameter, and is used in Tracor, OI, and Baseline instruments.
Model 103 has a 1.375" base and is used in HNU and SRI detectors.
Model 108-BTEX lamp’s higher output makes it ideal for detection of BTEX compounds.
BodyTextAccessories: PID lamp polishing kits contains an iron oxide cleaning compound, swabs, and instructions for use.
Customers Who Bought This Also Bought