High-Capacity Split Vent Trap, Restek
Fournisseur: Restek
Certificats
A propos de cet article
High-capacity split vent traps reduce the release of hazardous materials from the capillary split vent into the lab.
Traps are sold individually or in packs of five depending on model.
Delivery: Includes connecting lines and mounting kit
Customers Who Bought This Also Bought