Refillable Hydrocarbon Trap, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Hydrocarbon traps effectively remove trace impurities from carrier gas.
- Reduces organics to 0.1 ppm (assuming 100 ppm input)
- 60 µm frit prevents gas contamination by purifier particles
- Good for purge-and-trap systems
- Refillable and rechargeable
- Maximum operating pressure: 125 psig (862 kPa)
Refillable hydrocarbon traps feature a maximum flow of 5 L/min.
Spécifications
- Catalog No:
- RK22012
- RK22013
- Max. operating pressure:
- 17.2 bar (250 psi)
- 17.2 bar (250 psi)
- Maximum Flow Rate:
- 1.25 L/min. (0.33 gal./min.)
- 1.25 L/min. (0.33 gal./min.)
- Ø×L:
- 5.7×23.5 cm (21/4×91/4")
- 5.7×23.5 cm (21/4×91/4")