High-Capacity Moisture Trap, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Purged with ultra-high-purity helium; ready to use with any carrier gas including hydrogen.
- Reduces water to less than 15 ppb
- Maximum operating pressure: 250 psi (1.724 kPa)
- Maximum flow: 1.25 L/min
¹/₄" nickel-plated brass fittings.
Customers Who Bought This Also Bought