High-Capacity Oxygen Trap, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Removes up to 600 mg of oxygen or 2 g of water.
- Long life - typically purifies more than five 200 ft³ cylinders
- Reduces oxygen to 15 ppb
- Maximum operating pressure: 250 psi (1.724 kPa)
- Flow: 3 L/min @ 32 psi (221 kPa)
¹/₈" nickel-plated brass fittings.
Customers Who Bought This Also Bought