Injector Nut Kit for Shimadzu 17A, 2010, and 2014 GCs, Restek
Fournisseur: Restek
Certificats
A propos de cet article
For use with Shimadzu 17A, 2010, and 2014 GCs
- Can be Siltek® treated or untreated
Includes 17A injector nut, 0.4 mm graphite ferrule, and 1/16" stainless steel capillary nut.
Customers Who Bought This Also Bought