MSD Conversion Fitting, Restek
# de catalogue RK21314
Fournisseur: Restek
Spécifications
- Description:MSD Conversion Fitting
- Cat. no.:RK21314
- Supplier No.:21314
Spécifications
A propos de cet article
A flat, soft aluminum sealing ring deforms and butt-seals against the MSD interface.
- A standard Vespel® ferrule seals the column and 1/16" stainless steel nut
- Fitting is constructed of nickel-plated brass for longevity and softness
- Use any standard Vespel® or Vespel®/graphite 1/16" ferrule
- Improved design reduces chance of leaks
Ordering information: Includes a 1/16" stainless steel nut and two replacement sealing rings. Order ferrules separately.
Customers Who Bought This Also Bought