Standard Ferrules for ¹/₁₆, ¹/₈, and 1¹/₄" Fittings, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Available in Vespel®, graphite, or Vespel®/graphite material.
- Durable, leak-tight
- Preconditioned to eliminate out-gassing
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Recommended for mass spec transfer lines
Ferrules are available in a range of options to suit various sizings.
Customers Who Bought This Also Bought