Finger-Tight Capillary Column Nuts, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Rapidly tighten without wrenches; avoid stripped threads.
- Can be used with 0.25, 0.32, or 0.53 mm ID columns
Customers Who Bought This Also Bought
Rapidly tighten without wrenches; avoid stripped threads.