FID/Injector Cleaning Kit, Restek
# de catalogue RK20120
Fournisseur: Restek
Spécifications
- Description:FID/Injector Cleaning Kit
- Cat. no.:RK20120
- Supplier No.:20120
Spécifications
A propos de cet article
The FID/Injector cleaning kit is compatible with any injection port, allowing universal applicability.
- Stainless steel cleaning tools
Delivery: Kits include: Nylon tube brushes; pipe cleaner; stainless steel tube brushes; stainless steel surface brush; stainless steel jet reamers; and emery cloth.
Customers Who Bought This Also Bought