DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
# de catalogue RK21303
Fournisseur: Restek
Spécifications
- Description:DI Liner Adaptor for Agilent 5890 Packed Column GC
- Cat. no.:RK21303
- For Use With:for Agilent 5890 Packed Column GC
- Supplier No.:21303
Spécifications
A propos de cet article
Direct injection liner adapters use standard 1/16-inch capillary nuts and ferrules.
- Convenient wrench pad at base
- Includes 1/4-inch graphite ferrule and stainless steel nut
Use with Agilent or Restek DI glass inserts or Restek DI Uniliner® liners for an Agilent 5890 packed column GC.
Customers Who Bought This Also Bought