FID/NPD Adapter Fitting, Restek
# de catalogue 10855-866
Fournisseur: Restek
Spécifications
- Description:FID/NPD Adaptor Fitting
- Cat. no.:10855-866
- Supplier No.:20884
Spécifications
A propos de cet article
Includes 1/4 and 1/16" stainless steel nuts and 1/4" Vespel® and 0.4 mm ID graphite ferrules.
- Easy-to-use, sturdy, compact stainless steel fitting
- 1/16" nut uses standard graphite or Vespel®/graphite ferrules
- Wrench pad won’t turn when installing a capillary column
Customers Who Bought This Also Bought