Male Pipe to Valco® Internal Adapter, Restek
Fournisseur: Restek
Certificats
A propos de cet article
These adapters make a minimum-volume connection from a female pipe fitting on a pressure gauge or regulator to a Valco® zero-dead-volume fitting.
- Made of 300 series stainless steel; stainless steel ferrules included
Ideal for creating a zero-dead-volume fitting.
Customers Who Bought This Also Bought