Zero-Dead-Volume Valco® Internal Cross, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Use these cross connectors for making four-way connections with tubing.
- Connect four lines
- 300 series stainless steel; stainless steel ferrules included
- For 1/16" OD tubing
Designed so that no volume is introduced by the fitting which is not cleanly swept.
Customers Who Bought This Also Bought