Zero-Dead-Volume Valco® Internal Tee, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Use these tee connectors for making three-way connections with tubing.
- 300 series stainless steel; stainless steel ferrules included
- For 1/16" OD tubing
Designed so that no volume is introduced by the fitting which is not cleanly swept.
Customers Who Bought This Also Bought