Zero-Dead-Volume Valco® Internal Reducing Union, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Connect two sizes of tubing, using zero-dead-volume fittings on each end.
- For 1/8" to 1/16" OD tubing
- Stainless steel ferrules included
Designed so that no volume is introduced by the fitting which is not cleanly swept.
Customers Who Bought This Also Bought