Ferrule Removal Kit, Restek
Catalog # 89205-668
Supplier: Restek
Specifications
- Description:Ferrule Removal Kit
- Cat. no.:89205-668
- Supplier No.:20146
Specifications
About this item
The tapered tools in this kit have teeth designed to grip and remove fused silica adapter ferrules that have become stuck in the fitting detail.
Each kit has two tools: one for removing 1/32-inch adapter ferrules and one for removing 1/16-inch adapter ferrules.
Customers Who Bought This Also Bought