Open-End Wrenches, Restek
Catalog # RK20388
Supplier: Restek
Specifications
- Description:Open-End Wrenches, 1/4 – 3/16"
- Cat. no.:RK20388
- Supplier No.:20388
Specifications
About this item
High-quality miniature wrenches to use with MXT® low-dead-volume connectors.
- Durable fabrication
- Open-ended wrenches allow for easy application to fittings
- Range of sizings
Customers Who Bought This Also Bought