Replacement Ferrules for MXT® Connectors, ¹/₃₂" Stainless Steel, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
Replacement ferrules for MXT® connectors are constructed of stainless steel, sized to fit 1/32" fittings.
- Stainless steel fabrication
- 1/32" sizing
Customers Who Bought This Also Bought
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
45,90 $ to 336,74 $
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
81,56 $ to 628,06 $
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
149,40 $
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, Assembled, 9 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
152,62 $
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
148,39 $
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
1 853,47 $ to 1 853,47 $
SureSTART™ Glass Crimp Top Headspace Vials, 20 ml, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
3 538,41 $ to 3 538,41 $
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
113,85 $
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
178,08 $