Vespel®/Graphite Ferrule ¹/₃₂", Restek
Fournisseur: Restek
Certificats
A propos de cet article
Vespel®/Graphite ferrules are sized to fit 1/32" fittings.
Vespel®/Graphite hybrid material for excellent combination of sealing and workability.
Customers Who Bought This Also Bought