Replacement Nut, ¹/₃₂", Restek
# de catalogue RK20389
Fournisseur: Restek
Spécifications
- Description:1/32" Replacement Nut
- Cat. no.:RK20389
- Supplier No.:20389
Spécifications
A propos de cet article
1/32-inch replacement nuts are useful for complementing MXT® low-dead-volume connector kits.
- Durable
- Economical
- Compatible with all MXT® connector kits for metal columns
Convenient replacement part for use with MXT® connector kits.
Customers Who Bought This Also Bought