ContactUsLinkComponent
Replacement Nut, ¹/₃₂", Restek
Replacement Nut, ¹/₃₂", Restek
# de catalogue RK20389
Fournisseur:  Restek
1/32"Replacement Nut
Replacement Nut, ¹/₃₂", Restek
# de catalogue RK20389
Fournisseur:  Restek
Spécifications
  • Description:
    1/32" Replacement Nut
  • Cat. no.:
    RK20389
  • Supplier No.:
    20389
Spécifications
A propos de cet article
1/32-inch replacement nuts are useful for complementing MXT® low-dead-volume connector kits.
  • Durable
  • Economical
  • Compatible with all MXT® connector kits for metal columns
Convenient replacement part for use with MXT® connector kits.
Customers Who Bought This Also Bought