MXT® Low Dead Volume 'Y' Connector Kits for Metal Columns, Restek
Fournisseur: Restek
Certificats
A propos de cet article
MXT® low dead volume Y connector kits for metal columns can be used to connect a guard column or transfer line to an MXT® stainless steel column.
- Connect a guard column/transfer line to an MXT® stainless steel column
- Low thermal mass tracks rapid oven temperature programming
- Stainless steel ferrules and nuts
- Siltek® treatment ensures ultimate inertness
Connect two MXT® columns to one inlet or one MXT® column to two detectors. Each kit contains the MXT® union, three 1/32-inch ferrules and nuts.
Customers Who Bought This Also Bought