Blank Ferrules for ¹/₁₆" Fittings, Restek
# de catalogue RK20240
Fournisseur: Restek
Spécifications
- Description:Blank Ferrules—For 1/16-Inch Fittings
- Material:VG2, 60% Vespel/40% Graphite
- Cat. no.:RK20240
- Fitting Size:¹⁄₁₆"
- Supplier No.:20240
Spécifications
A propos de cet article
Blank ferrules have no hole, and are suited for 1/16" fittings.
- Stable to 400°C
- Seal with minimal torque
- Reusable
- Preferred for vacuum and high-pressure uses
VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability.
VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules.
Customers Who Bought This Also Bought