Mini Wool Puller/Inserter, Restek
# de catalogue RK20114
Fournisseur: Restek
Spécifications
- Description:Mini Wool Puller/Inserter
- Cat. no.:RK20114
- Supplier No.:20114
Spécifications
A propos de cet article
This miniature puller/inserter device is ideal for handily manipulating wool plugs.
- Slender design
- Easy to maneuver
Insert and remove wool plugs easily.
Caution: Not recommended for use with double gooseneck liners.
Customers Who Bought This Also Bought