Reducing Ferrules, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Reducing ferrules have a smoother surface and cleaner edges than conventional graphite ferrules.
- Preconditioned to eliminate outgassing
- High-purity, high-density graphite or Vespel®/graphite material
- Stable to 400°C
Available in graphite or Vespel®/graphite material.
Customers Who Bought This Also Bought