Two-Hole Ferrules for ¹/₈ and ¹/₁₆" Compression-Type Fittings, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Two-hole ferrules seal with minimal torque, are reusable, and are preferred for vacuum and high-pressure uses.
- Use 1/16-inch, two-hole ferrules with the 1/16-inch capillary inlet adaptor fitting kit
- Use 1/8-inch, two-hole ferrules with the 1/8-inch capillary inlet adaptor fitting kit
- Stable to 400°C
For use with 1/8" and 1/16" compression-type fittings.
Customers Who Bought This Also Bought