Capillary Ferrules for ¹/₁₆" Compression-Type Fittings, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Capillary ferrules are sized to fit 1/16" fittings.
- Available in Vespel®, graphite, or Vespel®/graphite material
For micropacked columns or 0.28 mm MXT columns.
Ordering information: For micropacked columns or 0.28 mm MXT columns.
Customers Who Bought This Also Bought