GRAPHPACK® 3D/2 Ferrules, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Gerstel GRAPHPACK® technology provides a complete system that quickly and reliable makes leak-tight, low-dead-volume connections.
- Ferrule internal diameter of 0.4 mm
Metal-jacketed graphite ferrules are the ideal seal for GC applications.
Customers Who Bought This Also Bought