Vu-Union Vespel®/Graphite Ferrules, Restek
Fournisseur: Avantor
Certificats
A propos de cet article
These ferrules have a maximum operating temperature of 400°C.
- Use with vacuum Vu-Union connectors
- 60% Vespel®/40% graphite
- 400°C maximum operating temperature
Customers Who Bought This Also Bought