Mininert® Precision Sampling Valves for Vials, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum.
- Screw-thread and crimp-top vial compatibility
- Variety of orifice sizes
Models are available for screw-cap and crimp-top vials. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial. Turn the threaded flange to secure a tight fit.
Customers Who Bought This Also Bought