Open-End Wrench Set, Restek
# de catalogue 10803-216
Fournisseur: Restek
Spécifications
- Description:Open-End Wrench Set
- Cat. no.:10803-216
- Supplier No.:20387
Spécifications
A propos de cet article
High-quality 1/4 x 5/16", 3/8 x 7/16", 7/16 x 1/2", and 1/2 x 9/16" wrenches for tightening a wide variety of chromatography fittings.
- Durable fabrication
- Open-ended wrenches allow for easy application to fittings
- Range of sizings
Customers Who Bought This Also Bought