Stainless Steel Jet Reamers/Ferrule Removers, Restek
# de catalogue RK20113
Fournisseur: Restek
Spécifications
- Description:Stainless Steel Jet Reamers/Ferrule Removers
- Cat. no.:RK20113
- Supplier No.:20113
Spécifications
A propos de cet article
Stainless steel jet reamers are great tools for cleaning detector jets or removing stuck ferrules from other small orifices.
- Useful for fine cleaning operations
- Slender design
Serrated design is optimal for removing silica deposits and other contaminants.
Customers Who Bought This Also Bought