ContactUsLinkComponent
Septum Puller, Restek
Septum Puller, Restek
# de catalogue RK20117
Fournisseur:  Restek
Septum Puller
Septum Puller, Restek
# de catalogue RK20117
Fournisseur:  Restek
Spécifications
  • Description:
    Septum Puller
  • Cat. no.:
    RK20117
  • Supplier No.:
    20117
Spécifications
A propos de cet article
Septum pullers lend an enhanced level of comfort in easily removing septa and O-rings from GC systems.
  • Effective at removing septa
  • Easily clear debris with fine pointed end
Use hooked end for removing septa and O-rings. The pointed end works well for removing stuck ferrules or debris.
Customers Who Bought This Also Bought