Open-End Wrenches, Restek
Fournisseur: Restek
Certificats
Certificats
A propos de cet article
High-quality wrenches for tightening capillary fittings.
- Durable construction from superior materials
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$73.75 to $568.62
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$568.25
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$43.20 to $44.79
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$184.08