Tapered Needle Files 4", Restek
# de catalogue RK20106
Fournisseur: Restek
Spécifications
- Description:4" Tapered Needle Files
- Cat. no.:RK20106
- Supplier No.:20106
Spécifications
A propos de cet article
Tapered needle files serve a variety of uses where fine points of manipulation are required.
- Very fine point
- Slender design
- Durable fabrication
These files can be used for many purposes. They are especially useful for removing ferrules that are lodged in injector or detector ports.
Customers Who Bought This Also Bought