Mini Hand Drill Set, Restek
# de catalogue RK20122
Fournisseur: Restek
Spécifications
- Description:Mini Hand Drill Set
- Cat. no.:RK20122
- Supplier No.:20122
Spécifications
A propos de cet article
Drill ferrules to the proper ID in seconds
- Compatible with a range of column internal diameters
- Multiple hand drills allow for precise drilling
Delivery: Includes three drills, for use with 0.25, 0.32, and 0.53 mm ID (0.4, 0.5, and 0.8 mm OD) capillary columns.
Customers Who Bought This Also Bought