ContactUsLinkComponent
Methane Cylinder Kit, Restek
Methane Cylinder Kit, Restek
# de catalogue RK20197
Fournisseur:  Restek
Methane Cylinder, Complete Kit
Methane Cylinder Kit, Restek
# de catalogue RK20197
Fournisseur:  Restek
Spécifications
  • Description:
    Methane Cylinder Kit
  • Cat. no.:
    RK20197
  • Supplier No.:
    20197
Spécifications
A propos de cet article
Setting the column flow rate by injecting methane and optimizing linear velocity is a preferred method for establishing reproducible retention times (ASTM Method E1510-93).
For optimizing linear velocity. The kit includes a Scotty® 14 cylinder containing 1% methane in helium, a MINICYL regulator, a syringe adapter, and a package of twenty-five septa for the adapter.
BodyTextAccessories: Replacement parts allow for continued maintenance of a methane cylinder kit, which is used for optimizing linear velocity.
Customers Who Bought This Also Bought