A propos de cet article
Measure dissolved oxygen levels without the difficulties of membrane replacement by using the Orion™ RDO™ Dissolved Oxygen Probe.
- Utilizes the latest lumiphore technology for measuring DO
- Once-a-year maintenance with a simple cap replacement
- Sensor cable lengths of 3, 6, 10, 15, or 30 m (10, 20, 30, 50, or 100')
- IP68-rated, waterproof
The Orion™ RDO™ dissolved oxygen probe offers a dissolved oxygen concentration range of 0.00 to 20.0mg/l, and a dissolved oxygen saturation range of 0.0 to 200%. Each probe order includes RDO probe, optical cap, stainless steel sensor guard, and calibration sleeve.
RDO (Rugged Dissolved Oxygen) Probe uses optical DO technology for problem-free measurement.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 97015-856
- 97015-860
- 97015-858
- Temperature range:
- 0 to 50°C
- 0 to 50°C
- 0 to 50°C