Princeton Review Coloring Books
# de catalogue 470101-562
Fournisseur: Random House
Spécifications
- Description:Biology Coloring Workbook
- Cat. no.:470101-562
- No. of Pages:352
- Length:21 cm
- Width:28 cm
- Supplier No.:0679778845
Spécifications
A propos de cet article
The Princeton Review Coloring Workbook Series uses a break-through approach to teaching these difficult concepts; engaging students in the learning process by ushering them through detailed diagrams and having them perform the seemingly simple task of coloring the structures as they locate and identify them.
Best of all, each workbook conforms to standard textbooks used in health and allied health curricula.
Customers Who Bought This Also Bought