Glas-Col® Flexible Fabric Heating Mantle, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes.
- Maximum temperature of 842°F (450°C)
- Require a controlled electrical power source to prevent overheatin
- Models for 120 VAC operation are CSA-certified
Hemispherical Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel supports (sold separately) are required to support mantle and allow attachment to ring stands. Supports have fabric strap basket to hold mantles;do not place mantle directly on ring stand.
Customers Who Bought This Also Bought