Ward's® Illite Shale
Catalog # 470025-406
Supplier: Ward's Science
Specifications
- Description:Illite Shale, 500 g
- Specimen Dimensions:Size will vary
- Cat. no.:470025-406
- Supplier No.:WARD470025-406
Specifications
About this item
Green shale which contains approximately 85% Illite. Impurities may include other clay minerals, such as micas and feldspars.
Customers Who Bought This Also Bought