Order Entry
ContactUsLinkComponent
Human Recombinant BMP2 (from E. coli)
Human Recombinant BMP2 (from E. coli)
# de catalogue 75794-174
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP2 (from E. coli)
# de catalogue 75794-174
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 μg
  • Tag sequence:
    His Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    Bone Morphogenetic Protein 2|BMP2
  • Protein/peptide name:
    BMP2
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    289-396
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
  • Cat. no.:
    75794-174
  • Supplier No.:
    90-374

Spécifications

A propos de cet article

Bone morphogenetic protein 2 (BMP-2) is a member of the BMP subgroup of the TGF-beta superfamily. It plays a dominant role in embryonic dorsalventral patterning, organogenesis, limb bud formation and bone formation and regeneration. BMP-2 signals through heterodimeric complexes composed of a type I receptor (Activin RI, BMP-RIA or BMP-RIB) and a type II receptor (BMP-RII or Activin RIIB). BMP-2 induces chondrocyte proliferation, endochondral bone formation, longitudinal bone growth and bone and cartilage repair. It induces ectopic bone formation or calcification by promoting osteogenic and chondrogenic differentiation in mesenchymal cells, stem cells and vascular smooth muscle cells. It also promotes the maintenance and repair of colonic epithelium, suppresses neuronal dopamine synthesis and release, induces apoptosis in medulloblastoma cells and is required for cardiac contractility.

  • Fusion-Tag: His Tag

This recombinant proteins is for research use only.

Customers Who Bought This Also Bought